Merck
  • A highly unusual palindromic transmembrane helical hairpin formed by SARS coronavirus E protein.
A highly unusual palindromic transmembrane helical hairpin formed by SARS coronavirus E protein.
Journal of molecular biology (2004-08-04)
Eyal Arbely, Ziad Khattari, Guillaume Brotons, Mutaz Akkawi, Tim Salditt, Isaiah T Arkin
ZUSAMMENFASSUNG
The agent responsible for the recent severe acute respiratory syndrome (SARS) outbreak is a previously unidentified coronavirus. While there is a wealth of epidemiological studies, little if any molecular characterization of SARS coronavirus (SCoV) proteins has been carried out. Here we describe the molecular characterization of SCoV E protein, a critical component of the virus responsible for virion envelope morphogenesis. We conclusively show that SCoV E protein contains an unusually short, palindromic transmembrane helical hairpin around a previously unidentified pseudo-center of symmetry, a structural feature which seems to be unique to SCoV. The hairpin deforms lipid bilayers by way of increasing their curvature, providing for the first time a molecular explanation of E protein's pivotal role in viral budding. The molecular understanding of this critical component of SCoV may represent the beginning of a concerted effort aimed at inhibiting its function, and consequently, viral infectivity.