Merck
  • Synthesis and characterization of linear aliphatic poly (methylene dicarboxylate) s from caesium dicarboxylates and bromochloromethane.
Synthesis and characterization of linear aliphatic poly (methylene dicarboxylate) s from caesium dicarboxylates and bromochloromethane.
Makromol. Chem. (1989)
Cimecioglu AL and East GC.