Merck
  • A1h NMR Pulse Gradient Spin-Echo (PGSE) Mass Transport Study of Dimethyl Oxalate and Ethylene Glycol: New Fuels for the DOFC.
A1h NMR Pulse Gradient Spin-Echo (PGSE) Mass Transport Study of Dimethyl Oxalate and Ethylene Glycol: New Fuels for the DOFC.
Electrochemical and Solid-State Letters (2000)
Suarez-Gustave S, et al.