Merck
  • Active sites in dibenzopyrenes: Synthesis and studies of 3-fluoro-and 2, 10-difluorobenzo (RST) pentaphene.
Active sites in dibenzopyrenes: Synthesis and studies of 3-fluoro-and 2, 10-difluorobenzo (RST) pentaphene.
Journal of Fluorine Chemistry (1976)
Boger E, et al.