Merck
  • Synthesis of poly (2-ethyl-2-oxazoline)-b-poly (styrene) copolymers via a dual initiator route combining cationic ring-opening polymerization and atom transfer radical polymerization.
Synthesis of poly (2-ethyl-2-oxazoline)-b-poly (styrene) copolymers via a dual initiator route combining cationic ring-opening polymerization and atom transfer radical polymerization.
Macromolecules (2008)
Becer CR, et al.