- The nitroimidazoles as radiosensitizers and cytotoxic agents.
The nitroimidazoles as radiosensitizers and cytotoxic agents.
The British journal of cancer. Supplement (1978-06-01)
E J Hall, R Miller, M Astro, F Rini
PMID277211
ZUSAMMENFASSUNG
Using hamster cells in culture, the radiosensitizing and cytotoxic properties of 8 electron-affinic drugs have been compared. These include nitrofurans derivatives as well as 2 and 5-nitroimidazoles. Most work has been performed with misonidazole for which it appears that, at 37 degree C, the concentration of drug required to produce a given level of cell killing is inversely proportional to the square of the exposure time. Misonidazole was also compared with X-rays for its ability to produce neoplastic transformations in vitro, using the C3H 10T 1/2 cell line.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
10T½, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.