Merck
  • Synthesis of Some New 2-(4-Methoxybenzothiazol-2'-yl amino)-4-(2-chloro-4-trifluoromethylanilino)-6-(substituted thioureido)-1, 3, 5-triazine as Antifungal Agents.
Synthesis of Some New 2-(4-Methoxybenzothiazol-2'-yl amino)-4-(2-chloro-4-trifluoromethylanilino)-6-(substituted thioureido)-1, 3, 5-triazine as Antifungal Agents.
Phosph. Sulfur Relat. Elem. (2009)
Sareen V, et al.