Merck
  • Effect of calcium and phytic acid on the activation of trypsinogen and the stability of trypsin.
Effect of calcium and phytic acid on the activation of trypsinogen and the stability of trypsin.
Journal of Agricultural and Food Chemistry (1992)
Caldwell RA