Merck
Neutral [2]rotaxane host systems that recognise halide anions in aqueous solvent mixtures.
Chemical communications (Cambridge, England) (2013-10-15)
James M Mercurio, Fergus Tyrrell, James Cookson, Paul D Beer
ZUSAMMENFASSUNG
Four pyridine N-oxide axle containing [2]rotaxanes have been synthesised via an anion templated threading-followed-by-stoppering strategy and shown to be the first examples of neutral interlocked host systems capable of recognising halide anions in aqueous solvent mixtures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin-N-oxid, 95%