Merck
  • Surface-enhanced Raman scattering of trans-1,2-bis (4-pyridyl)-ethylene on silver by theory calculations.
Surface-enhanced Raman scattering of trans-1,2-bis (4-pyridyl)-ethylene on silver by theory calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-18)
Zhiping Zhuang, Xiumin Shi, Yufeng Chen, Minghui Zuo
ZUSAMMENFASSUNG
Surface-enhanced Raman spectra of trans-1,2-bis (4-pyridyl)-ethylene (t-BPE) on silver foil were detected at laser line of 514.5, 633, 785 and 1064 nm, respectively. The structure of Ag-t-BPE, Ag4-t-BPE, Ag6-t-BPE, Ag10-t-BPE and Ag20-t-BPE complexes has been calculated using a local version of the Amsterdam density functional program package. The Raman spectra and electronic polarizability of t-BPE-Ag at 514.5, 633, 785 and 1064 nm excitation lines were calculated. The Raman bands of t-BPE were assigned according to the calculation of potential energy distribution. The experimental and calculated Raman spectra of t-BPE-Ag at 514.5, 633, 785 and 1064 nm were compared. The relative Raman intensities change at different excitation lines were discussed based on the Raman enhanced mechanism and surface selection rules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Silber, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95+%
Silber, foil, 2m coil, thickness 0.025mm, annealed, 99.95+%
Silber, foil, 100x100mm, thickness 1.0mm, as rolled, 99.95+%
Silber, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.95+%
Silber, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95+%
Silber, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Silber, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95+%
Silber, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95+%
Silber, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Silber, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%
Silber, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%
Silber, foil, 50x50mm, thickness 1.0mm, as rolled, 99.99+%
Silber, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95+%
Silber, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.95+%
Silber, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Silber, foil, 25mm disks, thickness 0.1mm, as rolled, 99.95+%
Silber, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95+%
Silber, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Silber, foil, 100x100mm, thickness 0.1mm, annealed, 99.95+%
Silber, foil, 100x100mm, thickness 0.20mm, as rolled, 99.95+%
Silber, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95+%
Silber, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Silber, foil, 6mm disks, thickness 0.003mm, 99.9%
Silber, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%
Silber, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.95+%
Silber, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Silber, foil, 0.5m coil, thickness 0.0125mm, 99.95+%
Silber, foil, 10mm disks, thickness 0.1mm, annealed, 99.95+%
Silber, foil, light tested, 150x150mm, thickness 0.075mm, as rolled, 99.95+%
Silber, foil, light tested, 25x25mm, thickness 0.0125mm, 99.95+%