Merck
  • A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-03-21)
Haimin Zhang, Yibing Li, Porun Liu, Ying Li, Dongjiang Yang, Huagui Yang, Huijun Zhao
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Zink, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95+%
Zink, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95+%
Zink, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95+%
Zink, foil, 1m coil, thickness 0.025mm, as rolled, 99.95+%
Zink, foil, 5m coil, thickness 0.125mm, as rolled, 99.95+%
Zink, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95+%
Zink, foil, 0.5m coil, thickness 0.05mm, as rolled, 99.95+%
Zink, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95+%
Zink, foil, 300x300mm, thickness 2.0mm, as rolled, 99.95+%
Zink, foil, 25x25mm, thickness 3mm, as rolled, 99.99+%
Zink, foil, 50x50mm, thickness 0.15mm, as rolled, 99.95+%
Zink, foil, 8mm disks, thickness 0.01mm, 99.9%
Zink, foil, 25x25mm, thickness 0.175mm, as rolled, 99.95+%
Zink, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Zink, foil, 150x150mm, thickness 0.175mm, as rolled, 99.95+%
Zink, foil, 100x100mm, thickness 0.175mm, as rolled, 99.95+%
Zink, foil, 100x100mm, thickness 3mm, as rolled, 99.99+%
Zink, foil, 5m coil, thickness 0.05mm, as rolled, 99.95+%
Zink, foil, 100x100mm, thickness 6mm, as rolled, 99.99+%
Zink, foil, 2m coil, thickness 0.05mm, as rolled, 99.95+%
Zink, foil, 15mm disks, thickness 0.01mm, 99.9%
Zink, foil, 8mm disks, thickness 0.02mm, 99.9%
Zink, foil, 10mm disks, thickness 0.01mm, 99.9%
Zink, foil, 4mm disks, thickness 0.01mm, 99.9%
Zink, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95+%
Zink, foil, 10mm disks, thickness 0.005mm, 99.9%
Zink, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95+%
Zink, foil, 300x300mm, thickness 0.35mm, as rolled, 99.95+%
Zink, foil, 50x50mm, thickness 0.175mm, as rolled, 99.95+%
Zink, foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%