Merck
Rib development in chick embryos analyzed by means of tantalum foil blocks.
The American journal of anatomy (1969-10-01)
R M Sweeney, R L Watterson
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Tantal, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Tantal, foil, 50x50mm, thickness 1.0mm, annealed, 99.9%
Tantal, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Tantal, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Tantal, foil, 150x150mm, thickness 0.15mm, annealed, 99.9%
Tantal, foil, 50x50mm, thickness 6.0mm, 99.9%
Tantal, foil, 25mm disks, thickness 0.015mm, 99.9%
Tantal, foil, light tested, 50x50mm, thickness 0.006mm, 99.9%
Tantal, foil, 25x25mm, thickness 0.5mm, annealed, 99.99%
Tantal, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Tantal, foil, 50x50mm, thickness 0.5mm, annealed, 99.9%
Tantal, foil, 8mm disks, thickness 0.034mm, as rolled, 99.9%
Tantal, foil, 8mm disks, thickness 0.25mm, annealed, 99.99%
Tantal, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Tantal, foil, not light tested, 100x100mm, thickness 0.01mm, 99.9%
Tantal, foil, 25x25mm, thickness 0.25mm, annealed, 99.9%
Tantal, foil, 10mm disks, thickness 0.100mm, annealed, 99.9%
Tantal, foil, 10mm disks, thickness 0.25mm, annealed, 99.9%
Tantal, foil, 10mm disks, thickness 0.5mm, annealed, 99.9%
Tantal, foil, 15mm disks, thickness 0.0075mm, 99.9%
Tantal, foil, 15mm disks, thickness 0.015mm, 99.9%
Tantal, foil, 15mm disks, thickness 0.034mm, as rolled, 99.9%
Tantal, foil, 15mm disks, thickness 0.100mm, annealed, 99.9%
Tantal, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
Tantal, foil, 6mm disks, thickness 0.009mm, 99.9%
Tantal, foil, 6mm disks, thickness 0.5mm, annealed, 99.9%
Tantal, foil, 8mm disks, thickness 0.025mm, as rolled, 99.9%
Tantal, foil, 8mm disks, thickness 0.125mm, annealed, 99.9%
Tantal, foil, light tested, 100x100mm, thickness 0.015mm, 99.9%
Tantal, foil, light tested, 100x100mm, thickness 0.034mm, as rolled, 99.9%