- Comparative study of the direct-lift and platinum foil techniques in the marginal discrepancy of collarless metal ceramic restorations.
Comparative study of the direct-lift and platinum foil techniques in the marginal discrepancy of collarless metal ceramic restorations.
Brazilian dental journal (1996-01-01)
W Matsumoto, R J Baez, H Panzeri
PMID9206363
ZUSAMMENFASSUNG
This study was carried out to evaluate the marginal discrepancy of collarless metal ceramic restorations, using a combination of three different techniques to manufacture the porcelain butt margin with two brands of body porcelain. Statistical analysis showed no significant difference between the techniques or brands of body porcelain used in this study.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Platin, foil, 25x25mm, thickness 0.1mm, as rolled, 99.99+%
Platin, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Platin, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Platin, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99.95%
Platin, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%
Platin, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Platin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Platin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Platin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Platin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Platin, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Platin, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
Platin, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95%
Platin, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95%
Platin, foil, not light tested, 25x25mm, thickness 0.0025mm, as rolled, 99.95%
Platin, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Platin, foil, 10mm disks, thickness 0.1mm, as rolled, 99.99+%
Platin, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Platin, foil, not light tested, 100x100mm, thickness 0.0025mm, as rolled, 99.95%
Platin, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99.95%
Platin, foil, 0.1m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platin, foil, 6mm disks, thickness 0.025mm, as rolled, 99.95%
Platin, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Platin, foil, 0.1m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Platin, foil, 1.2mm disks, thickness 0.025mm, as rolled, 99.95%
Platin, foil, 15mm disks, thickness 0.005mm, as rolled, 99.95%
Platin, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platin, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.999%
Platin, foil, 0.1m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%