Merck
Porcelain veneer fabrication. Platinum foil and refractory model techniques.
Ontario dentist (1994-05-01)
T Williams
ZUSAMMENFASSUNG
A quality porcelain veneer restoration can be made by either the refractory die method or by platinum foil. The platinum foil method produces excellent results with more traditional means. The refractory method can produce equally excellent results as well, but requires more effort in model making.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Platin, foil, 25x25mm, thickness 0.1mm, as rolled, 99.99+%
Platin, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Platin, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Platin, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99.95%
Platin, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%
Platin, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Platin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Platin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Platin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Platin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Platin, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Platin, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
Platin, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.95%
Platin, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95%
Platin, foil, not light tested, 25x25mm, thickness 0.0025mm, as rolled, 99.95%
Platin, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Platin, foil, 10mm disks, thickness 0.1mm, as rolled, 99.99+%
Platin, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Platin, foil, not light tested, 100x100mm, thickness 0.0025mm, as rolled, 99.95%
Platin, foil, not light tested, 25x25mm, thickness 0.01mm, as rolled, 99.95%
Platin, foil, 0.1m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platin, foil, 6mm disks, thickness 0.025mm, as rolled, 99.95%
Platin, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Platin, foil, 0.1m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Platin, foil, 1.2mm disks, thickness 0.025mm, as rolled, 99.95%
Platin, foil, 15mm disks, thickness 0.005mm, as rolled, 99.95%
Platin, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platin, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.999%
Platin, foil, 0.1m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%