Merck
  • Novel human BTB/POZ domain-containing zinc finger protein ZNF295 is directly associated with ZFP161.
Novel human BTB/POZ domain-containing zinc finger protein ZNF295 is directly associated with ZFP161.
Biochemical and biophysical research communications (2005-01-05)
Jun Wang, Jun Kudoh, Atsushi Takayanagi, Nobuyoshi Shimizu
ZUSAMMENFASSUNG
Human ZNF295 protein harbors a BTB/POZ domain and multiple krüppel (C(2)H(2)) type zinc finger domains, and thus belongs to a family of POK (POZ and krüppel) transcription factor. We have identified many transcript variants generated by the alternative splicing in 5' non-coding exons, an intra-exonic splicing in a coding region, and the use of three polyadenylation signals in the 3' UTR. The intra-exonic splicing removes 603-bp coding sequence, and thus ZNF295 gene produces two protein isoforms: ZNF295L with 1066 amino acid residues and ZNF295S with 865 amino acid residues, containing 9 and 5 zinc finger domains, respectively. ZNF295 is ubiquitously expressed in human fetal and adult tissues. Analysis of transcription activity of ZNF295 using various promoter-reporters demonstrated that ZNF295 acts as a transcription repressor, and contains two separate regions for repression activity: the BTB/POZ domain and the central region between BTB/POZ and ZF domains. Both ZNF295L and ZNF295S could interact not only with themselves and each other, but also with another POK protein ZFP161 known to function as a transcription repressor and an activator. We postulated that ZNF295 may be involved in the bi-directional control of gene expression in concert with ZFP161.