Merck
  • Functionalization of p-tert-butylcalix [6] arene by alkylation with 2-(chloromethyl) pyridine hydrochloride.
Functionalization of p-tert-butylcalix [6] arene by alkylation with 2-(chloromethyl) pyridine hydrochloride.
The Journal of Organic Chemistry (1993)
Neri P and Pappalardo S.