Merck
  • 411. Some derivatives of diphenylamine and a new synthesis of N-arylanthranilic acids and of acridones.
411. Some derivatives of diphenylamine and a new synthesis of N-arylanthranilic acids and of acridones.
Journal of the Chemical Society (1937)
Jamison MM and Turner EE.