Merck
  • Synthesis of [11-2H2], [8-2H2], [7-2H2], [6-2H2], [5-2H2], [4-2H2] and [3-2H2] cis-9-octadecenoates.
Synthesis of [11-<SUP>2</SUP>H<SUB>2</SUB>], [8-<SUP>2</SUP>H<SUB>2</SUB>], [7-<SUP>2</SUP>H<SUB>2</SUB>], [6-<SUP>2</SUP>H<SUB>2</SUB>], [5-<SUP>2</SUP>H<SUB>2</SUB>], [4-<SUP>2</SUP>H<SUB>2</SUB>] and [3-<SUP>2</SUP>H<SUB>2</SUB>] cis-9-octadecenoates.
Chemistry and Physics of Lipids (1979)
Tulloch AP.