Merck
  • Early depression of Ankrd2 and Csrp3 mRNAs in the polyribosomal and whole tissue fractions in skeletal muscle with decreased voluntary running.
Early depression of Ankrd2 and Csrp3 mRNAs in the polyribosomal and whole tissue fractions in skeletal muscle with decreased voluntary running.
Journal of applied physiology (Bethesda, Md. : 1985) (2012-01-28)
Michael D Roberts, Thomas E Childs, Jacob D Brown, J Wade Davis, Frank W Booth
ZUSAMMENFASSUNG
The wheel-lock (WL) model for depressed ambulatory activity in rats has shown metabolic maladies ensuing within 53-173 h after WL begins. We sought to determine if WL beginning after 21-23 days of voluntary running in growing female Wistar rats affected the mRNA profile in the polyribosomal fraction from plantaris muscle shortly following WL. In experiment 1, WL occurred at 0200 and muscles were harvested at 0700 daily at 5 h (WL5h, n = 4), 29 h (WL29h, n = 4), or 53 h (WL53h, n = 4) after WL. Affymetrix Rat Gene 1.0 ST Arrays were used to test the initial question as to whether WL affects mRNA occupancy on skeletal muscle polyribosomes. Using a false discovery rate of 15%, no changes in mRNAs in the polyribosomal fraction were observed at WL29h and eight mRNAs (of over 8,200 identified targets) were altered at WL53h compared with WL5h. Interestingly, two of the six downregulated genes included ankyrin repeat domain 2 (Ankrd2) and cysteine-rich protein 3/muscle LIM protein (Csrp3), both of which encode mechanical stretch sensors and RT-PCR verified their WL-induced decline. In experiment 2, whole muscle mRNA and protein levels were analyzed for Ankrd2 and Csrp3 from the muscles of WL5h (4 original samples + 2 new), WL29h (4 original), WL53h (4 original + 2 new), as well as WL173 h (n = 6 new) and animals that never ran (SED, 4-5 new). Relative to WL5h controls, whole tissue Ankrd2 and Csrp3 mRNAs were lower (P < 0.05) at WL53h, WL173h, and SED; Ankrd2 protein tended to decrease at WL53h (P = 0.054) and Csrp3 protein was less in WL173h and SED rats (P < 0.05). In summary, unique early declines in Ankrd2 and Csrp3 mRNAs were identified with removal of voluntary running, which was subsequently followed by declines in Csrp3 protein levels during longer periods of wheel lock.