Merck
  • Microchip-based synthesis and total analysis systems (?SYNTAS): chemical microprocessing for generation and analysis of compound libraries.
Microchip-based synthesis and total analysis systems (?SYNTAS): chemical microprocessing for generation and analysis of compound libraries.
Journal of the Chemical Society. Perkin Transactions 1 (2001)