- Hydrogen-bond-mediated cascade reaction involving chalcones: facile synthesis of enantioenriched trisubstituted tetrahydrothiophenes.
Hydrogen-bond-mediated cascade reaction involving chalcones: facile synthesis of enantioenriched trisubstituted tetrahydrothiophenes.
Organic letters (2012-02-09)
Jun-Bing Ling, Yu Su, Hai-Liang Zhu, Guan-Yu Wang, Peng-Fei Xu
PMID22313116
ZUSAMMENFASSUNG
A bifunctional squaramide catalyzed sulfa-Michael/aldol cascade reaction between 1,4-dithiane-2,5-diol and chalcones with a low catalyst loading has been developed. Trisubstituted tetrahydrothiophenes with three contiguous stereogenic centers are obtained in a highly stereocontrolled manner. Additionally, a remarkable temperature effect on reaction efficiency was observed and a synthetically potential gram-synthesis was also conducted.