Merck
  • Upstream stimulatory factors bind to insulin response sequence of the fatty acid synthase promoter. USF1 is regulated.
Upstream stimulatory factors bind to insulin response sequence of the fatty acid synthase promoter. USF1 is regulated.
The Journal of biological chemistry (1995-12-01)
D Wang, H S Sul
ZUSAMMENFASSUNG
Fatty acid synthase (FAS) plays a central role in de novo lipogenesis in mammals. The concentration or activity of FAS in liver and adipose tissue changes dramatically when animals are subjected to nutritional and hormonal manipulations. We previously reported that due to changes in transcription, FAS synthesis declines and increases in an insulin-dependent manner during fasting and refeeding, respectively, and that insulin administration of streptozotocin-diabetic mice stimulates FAS transcription. We previously mapped the FAS insulin response sequence (IRS) to the proximal promoter region from position -71 to position -50, which contains an E-box DNA binding motif. Here, using competition gel shift assays and specific upstream stimulatory factor (USF) antibodies, we identified USF1 and USF2 as major components of complexes that bind to the FAS IRS. UV-cross-linking experiments further supported that USFs bind the FAS IRS. We also found that the amount of the 43-kDa USF1 was dramatically increased in liver of refed rats. In contrast, the amount of USF2 remained the same in liver of fasted or refed rats. Moreover, a 17-kDa protein in both fasted and refed rat liver was recognized by anti-USF1 antibodies, and this 17-kDa USF1-related protein was expressed in a manner opposite to that of the 43-kDa USF1, i.e. high in liver of fasted rats and decreased in liver of refed rats. These data suggest that the regulation of USF expression may play an important role in the regulation of FAS transcription.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-USF1 (AB2) antibody produced in rabbit, IgG fraction of antiserum