- Cellular human CLE/C14orf166 protein interacts with influenza virus polymerase and is required for viral replication.
Cellular human CLE/C14orf166 protein interacts with influenza virus polymerase and is required for viral replication.
Journal of virology (2011-09-09)
Ariel Rodriguez, Alicia Pérez-González, Amelia Nieto
PMID21900157
ZUSAMMENFASSUNG
The influenza A virus polymerase associates with a number of cellular transcription-related factors, including RNA polymerase II. We previously described the interaction of influenza virus polymerase subunit PA with human CLE/C14orf166 protein (hCLE), a positive modulator of this cellular RNA polymerase. Here, we show that hCLE also interacts with the influenza virus polymerase complex and colocalizes with viral ribonucleoproteins. Silencing of hCLE causes reduction of viral polymerase activity, viral RNA transcription and replication, virus titer, and viral particle production. Altogether, these findings indicate that the cellular transcription factor hCLE is an important protein for influenza virus replication.