Merck
  • Photophysical properties of fullerenes and fullerene/N, N-diethylaniline charge-transfer complexes.
Photophysical properties of fullerenes and fullerene/N, N-diethylaniline charge-transfer complexes.
The Journal of Physical Chemistry (1992)
Wang Y.