Merck
  • Antimicrobial properties of the products from the reaction of various aminoalcohols and boric anhydride.
Antimicrobial properties of the products from the reaction of various aminoalcohols and boric anhydride.
Materials Chemistry and Physics (1988)
S Watanabe