Merck
  • Rapid synthesis of N, N'-disubstituted piperazines. Application to the preparation of No carrier added 1-(4-[18F] fluorophenyl) piperazine and of an [18F]-selective ligand of serotoninergic receptors (5HT2 antagonist).
Rapid synthesis of N, N'-disubstituted piperazines. Application to the preparation of No carrier added 1-(4-[18F] fluorophenyl) piperazine and of an [18F]-selective ligand of serotoninergic receptors (5HT2 antagonist).
Journal of the Chemical Society. Perkin Transactions 1 (1992)
Collins M, et al.