Merck
  • Synthesis of 5-pyridyl-2-furaldehydes via palladium-catalyzed cross-coupling with triorganozincates.
Synthesis of 5-pyridyl-2-furaldehydes via palladium-catalyzed cross-coupling with triorganozincates.
Organic letters (2002-02-01)
Donald R Gauthier, Ronald H Szumigala, Peter G Dormer, Joseph D Armstrong, R P Volante, Paul J Reider
ZUSAMMENFASSUNG
5-Pyridyl- and 5-aryl-2-furaldehydes are prepared from furaldehyde diethyl acetal in a four-step, one-pot procedure:(i) deprotonation; (2) Li to Zn transmetalation; (3) Pd-mediated cross-coupling; (4) aldehyde deprotection. Triorganozincate 7 was found to transfer all three groups in the Pd-catalyzed cross-coupling reaction with haloaromatics.