Merck
  • Surface-enhanced Raman spectroscopy of indole ring-terminated self-assembled monolayer on silver electrode.
Surface-enhanced Raman spectroscopy of indole ring-terminated self-assembled monolayer on silver electrode.
Chemija (2006)
Razmute I, et al.