Merck
  • Oxygen glucose deprivation causes mitochondrial dysfunction in cultivated rat hippocampal slices: protective effects of CsA, its immunosuppressive congener [D-Ser](8)CsA, the novel non-immunosuppressive cyclosporin derivative Cs9, and the NMDA receptor antagonist MK 801.
Oxygen glucose deprivation causes mitochondrial dysfunction in cultivated rat hippocampal slices: protective effects of CsA, its immunosuppressive congener [D-Ser](8)CsA, the novel non-immunosuppressive cyclosporin derivative Cs9, and the NMDA receptor antagonist MK 801.
Mitochondrion (2012-07-25)
Sonata Trumbeckaite, Zemfira Gizatullina, Odeta Arandarcikaite, Peter Röhnert, Stefan Vielhaber, Miroslav Malesevic, Gunter Fischer, Enn Seppet, Frank Striggow, Frank Norbert Gellerich
ZUSAMMENFASSUNG
We have introduced a sensitive method for studying oxygen/glucose deprivation (OGD)-induced mitochondrial alterations in homogenates of organotypic hippocampal slice cultures (slices) by high-resolution respirometry. Using this approach, we tested the neuroprotective potential of the novel non-immunosuppressive cyclosporin (CsA) derivative Cs9 in comparison with CsA, the immunosuppressive CsA analog [D-Ser](8)CsA, and MK 801, a N-methyl-d-aspartate (NMDA) receptor antagonist. OGD/reperfusion reduced the glutamate/malate dependent (and protein-related) state 3 respiration to 30% of its value under control conditions. All of the above drugs reversed this effect, with an increase to >88% of the value for control slices not exposed to OGD. We conclude that Cs9, [D-Ser](8)CsA, and MK 801, despite their different modes of action, protect mitochondria from OGD-induced damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(+)-MK-801 -hydrogen-maleat, powder