Merck
Drug distribution in wet granulation: foam versus spray.
Drug development and industrial pharmacy (2012-10-13)
Melvin X L Tan, Thanh H Nguyen, Karen P Hapgood
ZUSAMMENFASSUNG
Foam granulation technology is a new wet granulation approach for pharmaceutical formulations. This study evaluates the performance of foam and spray granulation in achieving uniform drug distribution using a model formulation. To observe wetting and nuclei formation, single drop/foam penetration experiments were performed on a static powder bed comprised of varying compositions of hydrophilic/hydrophobic glass ballotini, and hydrophilic lactose/hydrophobic salicylic acid respectively. High shear granulation experiments were performed in a 5L mixer using varying compositions of hydrophilic lactose and hydrophobic salicylic acid. Four percent hydroxylpropyl methylcellulose (HPMC) solution was delivered at 90 g/min as either a foam (92% FQ) or an atomized spray whilst recording impeller power consumption. After drying, the granule size distribution was measured and the granule composition was estimated using gravimetric filtration in methanol. Foam penetration was less dependent on the powder hydrophobicity compared to drop penetration. For glass ballotini powder mixtures, foam induced nucleation created nuclei with relatively uniform structure and size regardless of the powder hydrophobicity. For salicylic acid and lactose mixtures, increasing the proportion of salicylic acid reduced the nuclei granule size for both foam and drop binder addition. The granule drug distribution was not significantly affected by the binder addition method. Processing conditions, including liquid binder amount, impeller speed, wet massing, and the wettability properties of the formulation were the dominant factors for delivering homogeneous granules. The study reveals that foam and spray granulation involve different nucleation mechanisms - spray tends to incur early liquid penetration whereas foam granulation operates well in mechanical dispersion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylzellulose, viscosity: 4,000 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~86,000
Sigma-Aldrich
Methylzellulose, viscosity: 1,500 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Supelco
Lactose, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Sigma-Aldrich
Methylzellulose, viscosity: 15 cP
Sigma-Aldrich
Methylzellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
Methylzellulose, viscosity: 25 cP
Sigma-Aldrich
Methylzellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methylzellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~120,000
Sigma-Aldrich
Methocel® A15 LV, 27.5-31.5% methoxyl basis
Sigma-Aldrich
(Hydroxypropyl)methylcellulose, average Mn ~90,000
Lactose, wasserfrei, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methocel® MC, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methylzellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methylzellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP