Merck
High doses of cobalt induce optic and auditory neuropathy.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (2012-10-17)
Pietro Apostoli, Simona Catalani, Anna Zaghini, Andrea Mariotti, Pietro Luigi Poliani, Valentina Vielmi, Francesco Semeraro, Sarah Duse, Andrea Porzionato, Veronica Macchi, Alessandro Padovani, Maria Cristina Rizzetti, Raffaele De Caro
ZUSAMMENFASSUNG
The adverse biological effects of continuous exposure to cobalt and chromium have been well defined. In the past, this toxicity was largely an industrial issue concerning workers exposed in occupational setting. Nevertheless, recent reports have described a specific toxicity mediated by the high levels of cobalt and chromium released by metallic prostheses, particularly in patients who had received hip implants. Clinical symptoms, including blindness, deafness and peripheral neuropathy, suggest a specific neurotropism. However, little is known about the neuropathological basis of this process, and experimental evidence is still lacking. We have investigated this issue in an experimental setting using New Zealand White rabbits treated with repeated intravenous injections of cobalt and chromium, alone or in combination. No evident clinical or pathological alterations were associated after chromium administration alone, despite its high levels in blood and tissue while cobalt-chromium and cobalt-treated rabbits showed clinical signs indicative of auditory and optic system toxicity. On histopathological examination, the animals showed severe retinal and cochlear ganglion cell depletion along with optic nerve damage and loss of sensory cochlear hair cells. Interestingly, the severity of the alterations was related to dosages and time of exposure. These data confirmed our previous observation of severe auditory and optic nerve toxicity in patients exposed to an abnormal release of cobalt and chromium from damaged hip prostheses. Moreover, we have identified the major element mediating neurotoxicity to be cobalt, although the molecular mechanisms mediating this toxicity still have to be defined.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cobalt(II)-chlorid, 97%
Sigma-Aldrich
Cobalt(II)-chlorid, purum p.a., anhydrous, ≥98.0% (KT)
Sigma-Aldrich
Cobaltchlorid 0,1 mol/l -Lösung
Sigma-Aldrich
Chrom(II)-chlorid, 95%
Sigma-Aldrich
Chrom(II)-chlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Cobalt(II)-chlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Cobalt(II)-chlorid, AnhydroBeads, −10 mesh, 99.995% trace metals basis
Sigma-Aldrich
Chrom(II)-chlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis