Merck
  • Preconcentration and speciation of Cr(III) and Cr(VI) in water and soil samples by spectrometric detection via use of nanosized alumina-coated magnetite solid phase.
Preconcentration and speciation of Cr(III) and Cr(VI) in water and soil samples by spectrometric detection via use of nanosized alumina-coated magnetite solid phase.
Environmental monitoring and assessment (2013-02-23)
Hossein Tavallali, Gohar Deilamy-Rad, Pegah Peykarimah
ZUSAMMENFASSUNG
A novel nanomaterial has been developed for speciation of Cr(III) and Cr(VI) in water and soil samples. In this study, a new type of alumina-coated magnetite nanoparticles (Fe3O4/Al2O3 NPs) modified by the surfactant Triton X-114 has been successfully synthesized and used in magnetic mixed hemimicelles solid-phase extraction procedure. The procedure was based on the reaction of chromium(III) with 1-(2-pyridilazo)-2-naphtol as a ligand, yielding a complex, which was entrapped "in situ" in the surfactant hemimicelles. The concentration of chromium(III) was determined using flame atomic absorption spectrometry. After reduction of Cr(VI) to Cr(III) by ascorbic acid, the system was applied to the total chromium. Cr(VI) was then calculated as the difference between the total Cr and the Cr(III) content. This method can also be used for complicated matrices such as soil samples without any special pretreatment. Under the optimum conditions of parameters, the recoveries of Cr(III) by analyzing the spiked water and soil samples were between 98.6 and 100.8 % and between 96.5 and 100.7 %, respectively. Detection limits of Cr(III) were between 1.4 and 3.6 ng mL(-1) for water samples and 5.6 ng mg(-1) for soil samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumoxid, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminiumoxid, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminiumoxid, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminiumoxid, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminiumoxid, Corundum, α-phase, -100 mesh
Supelco
Aluminiumoxid, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminiumoxid, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminiumoxid, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis
Sigma-Aldrich
Aluminiumoxid, single crystal substrate, <0001>
Sigma-Aldrich
Aluminiumoxid, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminumoxid, mesoporös, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminiumoxid, for the determination of hydrocarbons