Merck
  • Mechanochemical activation of vincamine mediated by linear polymers: assessment of some "critical" steps.
Mechanochemical activation of vincamine mediated by linear polymers: assessment of some "critical" steps.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2013-03-26)
Dritan Hasa, Beatrice Perissutti, Mario Grassi, Michele R Chierotti, Roberto Gobetto, Valerio Ferrario, Davide Lenaz, Dario Voinovich
ZUSAMMENFASSUNG
The aim of the research was to investigate three "critical steps" that deserve particular attention during the mechanochemical activation of vincamine. The first step consisted in the selection of the best polymeric carrier/most affine stabiliser between linear PVP and NaCMC by using the GRID and the GRID based AutoDock software packages which permit to calculate their surface features and interactions. Moreover, the calculation of the partial and total solubility parameters supported the results obtained by GRID and AutoDock software. Then, after the selection of linear PVP-K30 as the suitable carrier, the influence of process and formulation variables on the amorphisation degree and solubility enhancement was studied, to select the most suitable process conditions and formulation parameters. Subsequently, the best performing samples were widely characterised using XRPD, TEM and SSNMR (including the proton relaxation ((1)H T1 NMR) time) techniques. These studies highlighted that all the coground samples were nanocrystalline solid dispersions indicating a dramatic difference between the amorphisation capacities of linear PVP-K30 and cross-linked PVP, used in previous analogous experiences. In particular, (13)C, (15)N and (1)H T1 NMR data point to a description of the system as a dispersion of nanocrystals in the polymer. In these dispersions vincamine is in a disordered crystalline state due to extensive interactions and contacts with PVP-K30 but the main hydrogen bonding motif characterising its packing remains. Again, differently from cross-linked PVP, dissolution studies revealed that linear PVP-K30 was able to promote a complete in vitro solubilisation of vincamine in some coground samples. What is more important, by using a linear polymer, drug-to-polymer and milling time variables appeared less influent on the solid state and in vitro properties of the composites. Finally, stability studies conducted for a period of 1year highlighted the high physical stability of the selected samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polyvinylpyrrolidon, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidon, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, Medium viscosity
Sigma-Aldrich
Polyvinylpyrrolidon, powder, average Mw ~55,000
Supelco
Poly-(vinylpolypyrrolidon), ~110 μm particle size
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, low viscosity
Sigma-Aldrich
Polyvinylpyrrolidon, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidon, average Mw ~1,300,000 by LS
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Polyvinylpyrrolidon, K 30
Sigma-Aldrich
Polyvinylpyrrolidon, K 90
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, High viscosity
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, viscosity 2500-6000 cP 
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Polyvinylpyrrolidon, powder, average Mw ~29,000
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Polyvinylpyrrolidon, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Natriumcarboxymethylcellulose Natriumsalz, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, medium viscosity
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Polyvinylpyrrolidon -Lösung, K 60, 45% in H2O
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose Natriumsalz, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Polyvinylpyrrolidon, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Supelco
Povidon, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyvinylpyrrolidon, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidon, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Povidon, meets USP testing specifications
Povidon, European Pharmacopoeia (EP) Reference Standard