Merck
Effect of sodium bicarbonate supplementation on 2000-m rowing performance.
International journal of sports physiology and performance (2013-04-13)
Ruth M Hobson, Roger C Harris, Dan Martin, Perry Smith, Ben Macklin, Kirsty J Elliott-Sale, Craig Sale
ZUSAMMENFASSUNG
The ability to buffer H+ could be vital to exercise performance, as high concentrations of H+ contribute to the development of fatigue. The authors examined the effect of sodium bicarbonate (SB) supplementation on 2000-m rowing-ergometer performance. Twenty male rowers (age 23 ± 4 y, height 1.85 ± 0.08 m, mass 82.5 ± 8.9 kg, 2000-m personal-best time 409 ± 16 s) completed two 2000-m rowing-ergometer time trials, separated by 48 h. Participants were supplemented before exercise with 0.3 g/kg body mass of SB or a placebo (maltodextrin; PLA). The trials were conducted using a double-blinded, randomized, counterbalanced crossover study design. Time to complete the 2000-m and time taken for each 500-m split were recorded. Blood lactate, bicarbonate, pH, and base excess were determined preexercise, immediately postexercise, and 5 min postexercise. Performance data were analyzed using paired t tests, as well as magnitude-based inferences; hematological data were analyzed using a repeated-measures ANOVA. Using paired t tests, there was no benefit of SB over PLA (P = .095). However, using magnitude-based inferences there was a likely beneficial effect of SB compared with PLA (PLA 412.0 ± 15.1 s, SB 410.7 ± 14.9 s). Furthermore, SB was 0.5 ± 1.2 s faster than PLA in the third 500 m (P = .035; possibly beneficial) and 1.1 ± 1.7 s faster in the fourth 500 m (P = .004; very likely beneficial). All hematological data were different between SB and PLA and were different from preexercise to postexercise. SB supplementation is likely to be beneficial to the performance of those competing in 2000-m rowing events, particularly in the second half of the event.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumbicarbonat -Lösung, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Supelco
Natriumbicarbonat-Konzentrat, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C