Merck
  • Gene expression responses for detecting sublethal effects of xenobiotics and whole effluents on a Xenopus laevis embryo assay.
Gene expression responses for detecting sublethal effects of xenobiotics and whole effluents on a Xenopus laevis embryo assay.
Environmental toxicology and chemistry (2013-05-03)
Laura San Segundo, Federica Martini, M Victoria Pablos
ZUSAMMENFASSUNG
In the present study, the authors investigated the effects of bisphenol A, chlorpyrifos, methylparaben, and 2 effluent samples from wastewater treatment plants located in the province of Madrid, Spain, on the messenger RNA expression of specific genes involved in early development (ESR1, pax6, bmp4, and myf5) and a gene involved in the general stress response (hsp70) during Xenopus laevis embryo development. Gene expression was analyzed after 4 h, 24 h, and 96 h of exposure by semiquantitative reverse-transcriptase-polymerase chain reaction. Concentration ranges of the compounds and dilutions for the samples were selected to cause morphological alterations in embryos after 96 h of exposure. Transcript levels of ESR1, pax6, and hsp70 were differentially altered at early developmental stages with patterns specific to the contaminant and the exposure time. However, further studies are needed to establish transcript levels of specific genes as biomarkers of sublethal effects in an environmental risk-assessment framework. Besides, studies including more generic responses, such as genes encoding antioxidant enzymes, together with genes related to embryonic development have to be developed to look for a battery of mechanistic endpoints for the evaluation of chemical exposure at the molecular level in a first-tier assessment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ReagentPlus®, ≥99.0%, crystalline
Supelco
Chlorpyriphos, PESTANAL®, analytical standard
Supelco
Methylparaben, PESTANAL®, analytical standard
Sigma-Aldrich
Methyl-4-hydroxybenzoat, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ≥99%, FCC
Supelco
Chlorpyriphos, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Methylparahydroxybenzoat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methyl-4-hydroxybenzoat, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl-4-hydroxybenzoat, analytical standard
Sigma-Aldrich
Methyl-4-hydroxybenzoat, tested according to Ph. Eur.