Merck
  • Investigation of alternative organic solvents and methods for the preparation of long-circulating and pH-sensitive liposomes containing cisplatin.
Investigation of alternative organic solvents and methods for the preparation of long-circulating and pH-sensitive liposomes containing cisplatin.
Journal of liposome research (2013-05-11)
Cristiane dos Santos Giuberti, Fernanda Alves Boratto, Ghania Degobert, Josianne Nicácio Silveira, Mônica Cristina Oliveira
ZUSAMMENFASSUNG
Recent studies using long-circulating and pH-sensitive liposomes containing cisplatin (SpHL-CDDP) have resulted in a formulation with improved pharmacokinetic, toxicity and tumor localization properties. In this study, SpHL-CDDP were prepared in both laboratory and pilot scales. This study evaluated the possibility of using the dehydration-rehydration method, as well as using alternative organic solvents (ethyl acetate/ethanol mixtures at 2:1 and 1:1 volume ratios), for the preparation of liposomes by the reverse-phase evaporation (REV) method. The influence of different concentrations of cisplatin (CDDP) (2.0, 1.0, 0.5 and 0.25 mg/mL) on the entrapment percentage and size of SpHL-CDDP was also investigated. In addition, carbohydrates were tested as cryoprotectants in a freeze-thaw study as a pretest to screen the type to be used in the freeze-drying process. A decrease in the encapsulation percentage of CDDP and an increase in the vesicle diameter could be observed for both liposome formulations prepared with ethyl acetate:ethanol mixtures, as compared with REV liposomes prepared with ethyl ether. It is important to note that after applying either quick or slow cooling, the mean diameter of SpHL (empty liposomes) proved to be similar when in the presence of cryoprotectants. In sum, the optimal processing conditions were achieved when using a 0.5 mg/mL CDDP solution, ethyl ether and the REV method, resulting in liposomal dispersions of mean diameters and homogeneities that were deemed suitable for intravenous administration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Supelco
Ethylacetat, analytical standard
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Supelco
Diethylether, analytical standard
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%