Merck
  • Improvement of cognitive function and physical activity of aging mice by human neural stem cells over-expressing choline acetyltransferase.
Improvement of cognitive function and physical activity of aging mice by human neural stem cells over-expressing choline acetyltransferase.
Neurobiology of aging (2013-06-05)
Dongsun Park, Yun-Hui Yang, Dae Kwon Bae, Sun Hee Lee, Goeun Yang, Jangbeen Kyung, Dajeong Kim, Ehn-Kyoung Choi, Seong Won Lee, Gon Hyung Kim, Jin Tae Hong, Kyung-Chul Choi, Hong Jun Lee, Seung U Kim, Yun-Bae Kim
ZUSAMMENFASSUNG
Aging is characterized by progressive loss of cognitive and memory functions as well as decrease in physical activities. In the present study, a human neural stem cell line (F3 NSC) over-expressing choline acetyltransferase (F3.ChAT), an enzyme responsible for acetylcholine synthesis, was generated and transplanted in the brain of 18-month-old male ICR mice. Four weeks post-transplantation, neurobehavioral functions, expression of ChAT enzyme, production of acetylcholine and neurotrophic factors, and expression of cholinergic nervous system markers in transplanted animals were investigated. F3.ChAT NSCs markedly improved both the cognitive function and physical activity of aging animals, in parallel with the elevation of brain acetylcholine level. Transplanted F3 and F3.ChAT cells were found to differentiate into neurons and astrocytes, and to produce ChAT proteins. Transplantation of the stem cells increased brain-derived neurotrophic factor (BDNF) and nerve growth factor (NGF), enhanced expression of Trk B, and restored host microtubule-associated protein 2 and cholinergic nervous system. The results demonstrate that human NSCs over-expressing ChAT improve cognitive function and physical activity of aging mice, not only by producing ACh directly but also by restoring cholinergic neuronal integrity, which might be mediated by neurotrophins BDNF and NGF.