Merck
Bothropic antivenom based on monoclonal antibodies, is it possible?
Toxicon : official journal of the International Society on Toxinology (2013-06-05)
Thiago S Frauches, Jorge H Petretski, Andrea C V Arnholdt, Elena B Lasunskaia, Eulógio C Q de Carvalho, Thereza L Kipnis, Wilmar D da Silva, Milton M Kanashiro
ZUSAMMENFASSUNG
Neutralizing monoclonal antibodies against three major toxic components of Bothrops atrox venom were produced and tested. The mAbs against phospholipase A2, hemorrhagic metalloprotease, and thrombin-like enzymes were produced in large amounts and purified with caprylic acid followed by ammonium sulfate precipitation. Purified mAbs were analyzed by SDS-PAGE and their ability to neutralize the respective toxins was tested. Five Swiss mice were injected i.p. with 13.5 mg of pooled mAbs and challenged via s.c. route with venom. Survival rate was recorded for the next 48 h. All mice treated and challenged with venom survived, whereas only one mouse in the control group survived. Bleeding time in mice treated with mAbs was similar to that observed in control mice. Our results show that monoclonal antibodies neutralized the lethal toxicity of Bothrops venom and indicate that there is a reasonable possibility of developing antivenoms based on humanized mAbs to treat victims of venomous animals in the future.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Octansäure, ≥99%
Sigma-Aldrich
Octansäure, ≥98%
Sigma-Aldrich
Ammoniumsulfat, BioXtra, ≥99.0%
Sigma-Aldrich
Ammoniumsulfat, BioUltra, ≥99.0% (T)
Supelco
Ammoniumsulfat, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammoniumsulfat, 99.999% trace metals basis
Sigma-Aldrich
Octansäure, ≥98%, FG
Sigma-Aldrich
Octansäure, natural, ≥98%, FG
Supelco
Octansäure, analytical standard
Supelco
Caprylsäure (Octansäure), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumsulfat, suitable for plant cell culture, ≥99.0%
Caprylsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammoniumsulfat, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium-14N2 Sulfat -Lösung, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N-sulfat -Lösung, 40 wt. % in H2O, 99.99 atom % 14N