Merck
  • Studies of asymmetric styrene cyclopropanation with a rhodium(II) metallopeptide catalyst developed with a high-throughput screen.
Studies of asymmetric styrene cyclopropanation with a rhodium(II) metallopeptide catalyst developed with a high-throughput screen.
Chirality (2013-06-12)
Ramya Sambasivan, Zachary T Ball
ZUSAMMENFASSUNG
Dirhodium metallopeptides have been developed as selective catalysts for asymmetric cyclopropanation reactions. A selective ligand sequence has been identified by screening on-bead metallopeptide libraries in a 96-well plate format. Efficient ligand synthesis and screening allows a 200-member library to be created and assayed in less than three weeks. These metallopeptides catalyze efficient cyclopropanation of aryldiazoacetates, providing asymmetric access to cyclopropane products in high diastereoselectivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Styrol, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
Styrol, analytical standard
Sigma-Aldrich
Rhodium, powder, 99.95% trace metals basis