Merck
  • Hordenine, a single compound produced during barley germination, inhibits melanogenesis in human melanocytes.
Hordenine, a single compound produced during barley germination, inhibits melanogenesis in human melanocytes.
Food chemistry (2013-06-19)
Sang-Cheol Kim, Jin-Hyunk Lee, Moo-Han Kim, Jung-A Lee, Yum Beom Kim, Eunsun Jung, Young-Soo Kim, Jongsung Lee, Deokhoon Park
ZUSAMMENFASSUNG
Melanin plays an important role protecting skin against ultraviolet light injury. However, increased production and accumulation of melanin results in a large number of skin disorders. Here, we identified hordenine as an active compound from germinated barley (Hordeum vulgare L.) and investigated the effects of hordenine on melanogenesis and its mechanisms of action in human epidermal melanocytes. We measured melanin content, tyrosinase activity, expression of melanogenesis-related proteins, and cAMP production. Melanin content was significantly inhibited by hordenine. The intracellular cAMP level was also reduced by hordenine. In addition, expression of microphthalmia-associated transcription factor (MITF), an upstream transcription factor of tyrosinase as well as tyrosinase, tyrosinase related protein (TRP)-1, and TRP-2, was inhibited by hordenine. Taken together, these results show that hordenine inhibited melanogenesis by suppressing cAMP production, which is involved in the expression of melanogenesis-related proteins and suggest that hordenine may be an effective inhibitor of hyperpigmentation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyramin, ≥98.0%
Sigma-Aldrich
Hordenin, ≥97.0% (HPLC)
Sigma-Aldrich
Tyramin, 98%, FG