Merck
  • Composite nanofibers doped with a phosphorescent Re(I) complex having restricted conjugation plane in its diamine ligand: preparation, characterization, photophysical property and oxygen-sensing performance.
Composite nanofibers doped with a phosphorescent Re(I) complex having restricted conjugation plane in its diamine ligand: preparation, characterization, photophysical property and oxygen-sensing performance.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-06-22)
Chen Lin, Wang Shaoyan, Wang Qi, Li Jing
ZUSAMMENFASSUNG
In this paper, we report a Re(I) complex of Re(CO)3(Cl-PYO)Br, where Cl-PYO stands for 2-(4-chlorophenyl)-5-(pyridin-2-yl)-1,3,4-oxadiazole, including its synthesis, identification, molecular structure, theoretical calculation and photophysical character. Re(CO)3(Cl-PYO)Br is found to be a yellow emitter with long excited state lifetime in pure N2 atmosphere. Theoretical calculation result suggests that this emission comes from a triplet metal-to-ligand-charge-transfer excited state. By doping Re(CO)3(Cl-PYO)Br into a polymer supporting matrix of poly(vinylpyrrolidone), the emission of the resulted composite materials is found to be sensitive towards various oxygen concentrations. The maximum sensitivity is obtained to be 7.88. Owing to the porous structure of fibrous poly(vinylpyrrolidone) matrix, a short response time of 11s towards molecular oxygen is also realized with high photostability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polyvinylpyrrolidon, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidon, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidon, powder, average Mw ~55,000
Supelco
Poly-(vinylpolypyrrolidon), ~110 μm particle size
Sigma-Aldrich
Polyvinylpyrrolidon, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidon, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidon, K 30
Sigma-Aldrich
Polyvinylpyrrolidon, K 90
Sigma-Aldrich
Polyvinylpyrrolidon, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidon, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Polyvinylpyrrolidon -Lösung, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidon, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Supelco
Povidon, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyvinylpyrrolidon, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Rhenium, powder, 99.995% trace metals basis
Sigma-Aldrich
Polyvinylpyrrolidon, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Povidon, meets USP testing specifications
Povidon, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis