Merck
  • Effect of alginate/nano-Ag coating on microbial and physicochemical characteristics of shiitake mushroom (Lentinus edodes) during cold storage.
Effect of alginate/nano-Ag coating on microbial and physicochemical characteristics of shiitake mushroom (Lentinus edodes) during cold storage.
Food chemistry (2013-06-26)
Tianjia Jiang, Lifang Feng, Yanbo Wang
ZUSAMMENFASSUNG
The effect of a novel alginate/nano-Ag coating material on the preservation quality of shiitake mushroom (Lentinus edodes) during 4±1°C storage was investigated. The results showed that the alginate/nano-Ag coating had quite a beneficial effect on the physicochemical and sensory quality, compared to the control treatment. After a 16-day storage, mushroom weight loss, softening, and browning of the alginate/nano-Ag coating were significantly inhibited. The lower microbial counts, including mesophilic, psychrophilic, pseudomonad, and yeasts and moulds, in treated mushrooms during storage should be attributed to the alginate/nano-Ag coating. Meanwhile, the contents of the reducing sugar, total sugar, total soluble solids and electrolyte leakage rate were increased to 3.9mg/g, 11.2mg/g, 5.1% and 16.5% for the alginate/nano-Ag coating and 3.7mg/g, 8.3mg/g, 6.3% and 31.7% for the control treatment. Therefore, the alginate/nano-Ag coating could be applied for preservation of the shiitake mushroom to expand its shelf life and improve its preservation quality.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumalginat, powder
Sigma-Aldrich
Natriumalginat
Sigma-Aldrich
Natriumalginat aus Braunalgen, Medium viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, low viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginsäure aus Braunalgen, powder
Sigma-Aldrich
Calciumalginat aus Braunalgen