Merck
Dysregulated heme oxygenase-ferritin system in pterygium pathogenesis.
Cornea (2013-06-25)
Timothy Fox, Katherine H Gotlinger, Michael W Dunn, Olivia L Lee, Tatyana Milman, Gerald Zaidman, Michal L Schwartzman, Lars Bellner
ZUSAMMENFASSUNG
Cyclooxygenase (COX)-, lipoxygenase (LOX)-, and cytochrome P450 monooxygenase (CYP)-derived eicosanoids have been implicated in ocular surface inflammation and neovascularization. These eicosanoids are subjected to regulation by enzymes, such as heme oxygenases (HOs) and ferritin. Quantitative polymerase chain reaction and lipidomics based on liquid chromatography-tandem mass spectrometry were performed on pterygia from patients undergoing surgical pterygium excision. Control tissues consisted of donor corneas. In addition, lipidomics based on liquid chromatography-tandem mass spectrometry was performed on tears collected from patients before the surgery. Messenger RNA (mRNA) expression of HO-2, the constitutive HO isoform, was upregulated by 40% in pterygia compared with control tissue, whereas the mRNA level of the inducible form, HO-1, was downregulated by more than 50%. Levels of CYP4B1 mRNA showed an approximate 2-fold increase in pterygia compared with control. Lipidomic analysis of tissues indicated a moderate elevation in Prostaglandin E2 and thromboxane B2 levels in pterygia compared with control. Among the LOX-derived metabolites, the antiinflammatory-hydroxyeicosatetraenoic acid (15-HETE) levels were significantly reduced in pterygia (79.3 ± 48.11 pg/mg protein) compared with control (586.2 ± 213.5 pg/mg protein), whereas the proinflammatory LOX- and CYP4B1-derived 12-HETE levels were 10-fold higher in pterygia (2768 ± 832.3 pg/mg protein) compared with control (231.4 ± 87.35 pg/mg protein). Prostaglandin E2 and HETEs were also present in tears from patients with pterygium but were not detected in tears from healthy volunteers. The mRNA expression levels of both light and heavy chain ferritin were 60% and 30% lower, respectively, in pterygia compared with control. We believe that a dysfunctional HO-ferritin system leads to increased levels of proinflammatory mediators, thus contributing to the inflammation characteristic of pterygia.