Merck
  • Bronchial artery embolization to control hemoptysis: comparison of N-butyl-2-cyanoacrylate and polyvinyl alcohol particles.
Bronchial artery embolization to control hemoptysis: comparison of N-butyl-2-cyanoacrylate and polyvinyl alcohol particles.
Radiology (2013-06-27)
Sungmin Woo, Chang Jin Yoon, Jin Wook Chung, Sung-Gwon Kang, Hwan Jun Jae, Hyo-Cheol Kim, Nak Jong Seong, Young-Joo Kim, Young-Nam Woo
ZUSAMMENFASSUNG
To retrospectively compare safety and effectiveness of embolic agents polyvinyl alcohol (PVA) particles versus n-butyl-2-cyanoacrylate (NBCA) for bronchial artery embolization (BAE) for control of hemoptysis. Institutional review board approved this retrospective study; informed consent was waived. From January 2005 to December 2008, 406 patients (242 men, 164 women; age range, 6-92 years) with major hemoptysis underwent BAE by using PVA particles (n = 293) or NBCA (n = 113). Technical and clinical success, complications, hemoptysis-free survival rates, and causes of recurrent hemoptysis were compared between PVA and NBCA groups. The differences in hemoptysis-free survival rates were assessed between subgroups stratified to underlying diseases. The predictive factor for recurrent hemoptysis was identified with Cox proportional hazard regression model. Technical success was achieved in 93.9% (275 of 293) and 96.5% (109 of 113) of patients for PVA and NBCA, respectively (P = .463); clinical success was achieved in 92.2% (270 of 293) and 96.5% (109 of 113) of patients for PVA and NBCA, respectively (P = .180). Overall and major complication rates were not statistically different (overall complication rates: 34.1% for PVA, 31.0% for NBCA; P = .56; major complication rates: 0.3% for PVA, 0% for NBCA; P > .999). The 1-, 3-, and 5-year hemoptysis-free survival rates were, respectively, 77%, 68%, and 66% for PVA and 88%, 85%, and 83% for NBCA (P = .01). Recanalization of previously embolized vessels was more frequent in PVA group (21.5%) than in NBCA group (1.8%; P < .001). NBCA group showed hemoptysis-free survival rates superior to PVA group in patients with bronchiectasis (P = .016). PVA (P = .050) and aspergilloma (P < .001) were predictive factors for recurrent hemoptysis. BAE with NBCA provided higher hemoptysis-free survival rates compared with PVA particles without increasing complication rates. This improvement was evident in patients with bronchiectasis and was caused by more durable embolic effect than PVA particles. http://radiology.rsna.org/lookup/suppl/doi:10.1148/radiol.13130046/-/DC1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(vinylalkohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 150,000 (average), 99+% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinylalkohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinylalkohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Poly(vinylalkohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Kuraray Poval® 56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000