Merck
  • Improved tolerance of recombinant Escherichia coli to the toxicity of crude glycerol by overexpressing trehalose biosynthetic genes (otsBA) for the production of β-carotene.
Improved tolerance of recombinant Escherichia coli to the toxicity of crude glycerol by overexpressing trehalose biosynthetic genes (otsBA) for the production of β-carotene.
Bioresource technology (2013-07-09)
Anh Do Quynh Nguyen, You Geun Kim, Sung Bae Kim, Chang-Joon Kim
ZUSAMMENFASSUNG
This study aims to investigate whether overexpressing the trehalose biosynthetic gene, otsBA operon, in β-carotene-producing recombinant Escherichia coli protects cells from toxic impurities in crude glycerol. The concentrations of potassium and methanol in crude glycerol were too low to inhibit cell growth. Cell growth and production in control cell culture were inhibited significantly in the presence of a small amount of crude fatty acids. Peroxides were generated in the presence of crude fatty acids during autoclaving and, thus, the inhibitory effect of crude fatty acids was caused primarily by these peroxides. Engineered cells overexpressing otsBA tolerated crude fatty acids (≤42 wet-g/L), methanol (≤7.5 g/L), and t-BuOOH (≤60 μM) in separate experiments and tolerated up to 60 g/L crude glycerol. These results demonstrate that overexpressing otsBA endowed cells with the capacity to tolerate the toxicity of crude glycerol for direct use.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Carotin, synthetic, ≥95% (HPLC), crystalline
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
β-Carotin, synthetic, ≥93% (UV), powder
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
β-Carotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous