Merck
Comparison of ionic and non-ionic drug release from multi-membrane spherical aerogels.
International journal of pharmaceutics (2013-07-16)
Anja Veronovski, Zeljko Knez, Zoran Novak
ZUSAMMENFASSUNG
The presented research was oriented towards the preparation of dry biodegradable alginate aerogels with multi-membranes using a multi-step sol-gel process with potential applications as carriers during oral drug delivery. First alginate spherical hydrogels were formed in CaCl2 or BaCl2 solutions by ionic cross-linking. These cores were further immersed into alginate sodium solution, filtered through a sieve, and dropped into the salt solution again. Multi-membrane hydrogels were obtained by repeating the above process. They were further converted into aerogels by supercritical drying. The effect of the number of membranes was investigated regarding the loading and release of the model drugs nicotinic acid and theophylline. Moreover, the efficiencies of Ba(2+) and Ca(2+) metal ions for forming tridimensional networks that retain and extend drug release were also investigated. Nicotinic acid release was prolonged by adding membranes around the core and using Ca(2+) for cross-linking. However, retarded theophylline release was only obtained by using Ba(2+) for cross-linking. Namely, by increasing the number of membranes and BaCl2 concentration drug release became linear versus time in all studied cases. In the case of nicotinic acid loading increased by adding membranes around the core, however, for theophylline the opposite results were obtained due to the different nature of the model drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calciumchlorid Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Calciumchlorid Dihydrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Bariumchlorid Dihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Natriumalginat, powder
Sigma-Aldrich
Calciumchlorid Dihydrat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumalginat
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calciumchlorid Dihydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Bariumchlorid Dihydrat, ≥99%
Sigma-Aldrich
Theophyllin, anhydrous, ≥99%, powder
Sigma-Aldrich
Natriumalginat aus Braunalgen, Medium viscosity
Sigma-Aldrich
Bariumchlorid, 99.9% trace metals basis
Sigma-Aldrich
Natriumalginat aus Braunalgen, low viscosity
Sigma-Aldrich
Nicotinsäure, ≥99.5% (HPLC)
Sigma-Aldrich
Natriumalginat aus Braunalgen, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Nicotinsäure, ≥98%
Supelco
Nicotinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Alginsäure aus Braunalgen, powder
Supelco
Nicotinsäure, analytical standard
Sigma-Aldrich
Bariumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Nicotinsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Calciumchlorid Dihydrat, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calciumchlorid Dihydrat, tested according to Ph. Eur.
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat, BioXtra, ≥99.0%
Supelco
Theophyllin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bariumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis