Merck
  • A sensitive LC-MS/MS method for simultaneous determination of six flavonoids in rat plasma: application to a pharmacokinetic study of total flavonoids from mulberry leaves.
A sensitive LC-MS/MS method for simultaneous determination of six flavonoids in rat plasma: application to a pharmacokinetic study of total flavonoids from mulberry leaves.
Journal of pharmaceutical and biomedical analysis (2013-07-16)
Jun He, Ying Feng, Hui-Zi Ouyang, Bin Yu, Yan-Xu Chang, Gui-Xiang Pan, Gai-Ying Dong, Tao Wang, Xiu-Mei Gao
ZUSAMMENFASSUNG
A simple and sensitive LC-MS/MS method has been developed and validated for the determination of rutin, isoquercitrin, astragalin, quercetin, kaempferol and isorhamnetin in rat plasma using naringin as the internal standard (IS). The plasma samples were pretreated and extracted by liquid-liquid extraction. Chromatographic separation was accomplished on a C18 column with a 10 min gradient elution using acetonitrile and 0.1% formic acid aqueous solution as mobile phase at a flow rate of 0.3 mL min(-1). A tandem mass spectrometric detection was conducted using multiple reaction monitoring (MRM) via an electrospray ionization (ESI) source and operating in the negative ionization mode. The lower limit of quantitation (LLOQ) of each analyte was lower than 1 ng mL(-1). Intra-day and inter-day precisions were less than 11.9%. The relative errors of accuracy were in the range of -9.2% to 6.1%. The mean recoveries of flavonoids and IS were higher than 53.8%. The proposed method was further applied to investigate the pharmacokinetics of all analytes after a single oral administration of total flavonoids from mulberry leaves to rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isorhamnetin, ≥95.0% (HPLC)
Sigma-Aldrich
Kaempferol-3-β-D-glucopyranosid, ≥97.0% (HPLC)
Supelco
Isorhamnetin, analytical standard
Isoquercitrosid, European Pharmacopoeia (EP) Reference Standard