Merck
  • Gelatine enhances drug dispersion in alginate bilayer film via the formation of crystalline microaggregates.
Gelatine enhances drug dispersion in alginate bilayer film via the formation of crystalline microaggregates.
International journal of pharmaceutics (2013-07-17)
Hnin-Ei Thu, Shiow-Fern Ng
ZUSAMMENFASSUNG
In our previous study, a novel alginate-based bilayer film for slow-release wound dressings was successfully developed. We found that alginate alone yielded poor films; however, the addition of gelatine had significantly enhanced the drug dispersion as well as the physical properties. Here, an investigation of the drug-polymer interactions in the bilayer films was carried out. Drug content uniformity test and microscopy observation revealed that the addition of gelatine generated bilayer films with a homogenous drug distribution within the matrix. The FTIR and XRD data showed an increase in film crystallinity which might infer the presence of drug-polymer crystalline microaggregates in the films. DSC confirmed the drug-polymer interaction and indicated that the gelatine has no effect on the thermal behaviour of the microaggregates, suggesting the compatibility of the drug and excipients in the bilayer films. In conclusion, the addition of gelatine can promote homogenous dispersion of hydrophobic drugs in alginate films possibly through the formation of crystalline microaggregates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumalginat, powder
Sigma-Aldrich
Natriumalginat
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumalginat aus Braunalgen, Medium viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, low viscosity
Sigma-Aldrich
Natriumalginat aus Braunalgen, BioReagent, suitable for immobilization of micro-organisms
Sigma-Aldrich
Alginsäure aus Braunalgen, powder
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Ibuprofen für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen
Supelco
Ibuprofen Natriumsalz, ≥98% (GC)
Sigma-Aldrich
Calciumalginat aus Braunalgen
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ibuprofen, meets USP testing specifications