Merck
  • Determining if low dose hyper-radiosensitivity (HRS) can be exploited to provide a therapeutic advantage: a cell line study in four glioblastoma multiforme (GBM) cell lines.
Determining if low dose hyper-radiosensitivity (HRS) can be exploited to provide a therapeutic advantage: a cell line study in four glioblastoma multiforme (GBM) cell lines.
International journal of radiation biology (2013-07-19)
Diane Schoenherr, Sarah A Krueger, Lynn Martin, Laure Marignol, George D Wilson, Brian Marples
ZUSAMMENFASSUNG
To determine if ultra-fractionation using repeated pulses of radiation (10 × 0.2 Gray [Gy]) would be more cytotoxic than continuously-delivered radiation to the same total dose (2 Gy) in four glioma cell lines. Human T98G, U373, U87MG and U138MG cells were conventionally X-irradiated with 0.1-8 Gy and clonogenic survival assessed. Next, cells were treated with either a single dose of 2 Gy or 10 pulses of 0.2 Gy using a 3-min inter-pulse interval and DNA (Deoxyribonucleic acid) repair (pHistone H2A.X), G2-phase cell cycle checkpoint arrest (pHistone H3) and apoptosis (caspase-3) compared between the two regimens. A dose of 0.2 Gy was selected as this reflects the hyper- radiosensitivity (HRS)/increased radioresistance (IRR) transition point of the low-dose cell survival curve. T98G, U87MG and U138MG exhibited distinct HRS responses and survival curves were well-described by the Induced Repair model. Despite the prolonged delivery time, ultra-fractionation (10 × 0.2 Gy) was equally effective as a single continuously-delivered 2 Gy dose. However, ultra-fractionation was more effective when given for five consecutive days to a total dose of 10 Gy. The increased effectiveness of ultra-fractionation could not be attributed directly to differences in DNA damage, repair processes or radiation-induced apoptosis. Ultra-fractionation (10 × 0.2 Gy) is an effective modality for killing glioma cell lines compared with standard 2 Gy dosing when multiple days of treatment are given.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)