Merck
Efficient p-type dye-sensitized solar cells based on disulfide/thiolate electrolytes.
Nanoscale (2013-07-19)
Xiaobao Xu, Bingyan Zhang, Jin Cui, Dehua Xiong, Yan Shen, Wei Chen, Licheng Sun, Yibing Cheng, Mingkui Wang
ZUSAMMENFASSUNG
Herein, an organic redox couple 1-methy-1H-tetrazole-5-thiolate (T(-)) and its disulfide dimer (T2) redox shuttle, as an electrolyte, is introduced in a p-type dye-sensitized solar cell (DSC) on the basis of an organic dye (P1) sensitizer and nanocrystal CuCrO2 electrode. Using this iodide-free transparent redox electrolyte in conjunction with the sensitized heterojunction, we achieve a high open-circuit voltage of over 300 mV. An optimal efficiency of 0.23% is obtained using a CoS counter electrode and an optimized electrolyte composition under AM 1.5 G 100 mW cm(-2) light illumination which, to the best of our knowledge, represents the highest efficiency that has so far been reported for p-type DSCs using organic redox couples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrazol -Lösung, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile